Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Allolithocholic acid | 2276-94-0 | MFCD00013892 | 99.7% | C24H40O3 | 10MG
SDP

Catalog No. 5000258322
Encompass_Preferred

Allolithocholic acid is an endogenous steroid bile acid used in biochemical and metabolic research to study bile acid metabolism and lipid transport. It facilitates excretion, absorption, and transport of fats and sterols in the intestine and liver. The product is supplied as a high-purity solid with specified solubility and storage recommendations for both in vitro and in vivo preparations.

  • Endogenous steroid bile acid relevant to bile acid metabolism studies.
  • High purity (99.69%) suitable for analytical and biological assays.
  • Available in multiple sizes to support small- and medium-scale experiments.
  • Soluble in DMSO (25 mg/mL) with ultrasonic assistance; handle hygroscopic solvent accordingly.
  • Includes recommended in vivo dissolution protocols for formulation development.
  • Storage recommendations provided for solid and solution states to maintain stability.

Catalog No. 50-002-58322 Supplier Medchemexpress LLC Supplier No. HY14371210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Allolithocholic acid is an endogenous steroid bile acid used in biochemical and metabolic research to study bile acid metabolism and lipid transport. It facilitates excretion, absorption, and transport of fats and sterols in the intestine and liver. The product is supplied as a high-purity solid with specified solubility and storage recommendations for both in vitro and in vivo preparations.

  • Endogenous steroid bile acid relevant to bile acid metabolism studies.
  • High purity (99.69%) suitable for analytical and biological assays.
  • Available in multiple sizes to support small- and medium-scale experiments.
  • Soluble in DMSO (25 mg/mL) with ultrasonic assistance; handle hygroscopic solvent accordingly.
  • Includes recommended in vivo dissolution protocols for formulation development.
  • Storage recommendations provided for solid and solution states to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.