Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allopregnanolone, ≥95%, MP Biomedicals™

Catalog No. ICN19588605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19588605 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19588605 Supplier MP Biomedicals, Inc Supplier No. 0219588605

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 516-54-1
Molecular Formula C21H34O2
Molecular Weight (g/mol) 318.50
MDL Number MFCD00003677
InChI Key AURFZBICLPNKBZ-SYBPFIFISA-N
Synonym allopregnanolone, allopregnan-3alpha-ol-20-one, allotetrahydroprogesterone, 3alpha-hydroxy-5alpha-pregnan-20-one, brexanolone, 3a-hydroxy-5a-pregnan-20-one, unii-s39xz5qv8y, 3-alpha,5-alpha-pregnanolone, 3alpha-oh dhp, 5alpha-pregnan-3alpha-ol-20-one
PubChem CID 92786
ChEBI CHEBI:50169
IUPAC Name 1-[(3R,5S,8R,9S,10S,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Color White
Quantity 5 mg
Formula Weight 318.5
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Allopregnanolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.