Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Allopregnanolone ELISA Kit

Catalog No. EIAALLOX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAALLOX10 10 x 96 Tests
EIAALLO 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIAALLOX10 Supplier Invitrogen™ Supplier No. EIAALLOX10
Add to Cart
Add to Cart
Request a Quote

ELISA

The Allopregnanolone Competitive ELISA quantitates allopregnanolone in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.∣Principle of the method: The Progesterone Metabolites Competitive ELISA research-use-only kit is designed to quantitatively measure allopregnanolone independent of species. A allopregnanolone standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit allopregnanolone antibodies and allopregnanolone-peroxidase conjugate is added to the wells. The allopregnanolone-peroxidase conjugate and any allopregnanolone in the sample will compete to bind to the anti-rabbit allopregnanolone antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound allopregnanolone-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of allopregnanolone present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number 3 alpha,5 alpha-tetrahydroprogesterone; 3 alpha-hydroxy-5 alpha-pregnan-20-one; allopregnanolone; Brexanolone; C21H34O2; Zulresso
Assay Range 156.25 to 10,000 pg/mL
Assay Sensitivity 129.7 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Sufficient For 10 x 96 Assays
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 8%
Intraassay CV 7
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Storage Requirements -20°C
Target Allopregnanolone
Test Time 45 mins
Total Assay Time 16 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.