Learn More
U.S. Pharmacopeia Allopurinol, 315-30-0, MFCD00599413, 250 mg
Catalog No. NC0994783
Encompass
Empirical Formula (Hill Notation): C5H4N4O, Molecular Weight: 136.11, mp: >300 °C, Synonym: 1H-Pyrazolo(3,4-d)pyrimidin-4-ol, 4-Hydroxypyrazolo(3,4-d)pyrimidine, 4-Hydroxypyrazolo[3,4-d]pyrimidine, HPP.
Catalog No. NC0994783 Supplier U.S. Pharmacopeia Supplier No. 1013002
Empirical Formula (Hill Notation): C5H4N4O, Molecular Weight: 136.11, mp: >300 °C, Synonym: 1H-Pyrazolo(3,4-d)pyrimidin-4-ol, 4-Hydroxypyrazolo(3,4-d)pyrimidine, 4-Hydroxypyrazolo[3,4-d]pyrimidine, HPP.