Learn More
Selleck Chemical LLC Allopurinol, 25mg | CAS No. 315-30-0 | Purity: 99.98%
SDP
Catalog No. NC3662304
Encompass_Preferred
Allopurinol is a purine analog inhibitor of the enzyme xanthine oxidase, used to treat gout or kidney stones, and to decrease levels of uric acid.
Catalog No. NC3662304 Supplier Selleck Chemical LLC Supplier No. S163025MG
Allopurinol is a purine analog inhibitor of the enzyme xanthine oxidase, used to treat gout or kidney stones, and to decrease levels of uric acid.