Learn More
Medchemexpress LLC Allopurinol | 315-30-0 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB021910MM/1ML
Encompass_Preferred
Allopurinol is a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. It can be used in the research of hyperuricemia and gout. It decreases the expression of HIF-1α and HIF-2α protein. It also shows anti-depressant and anti-nociception activity, and anti-leishmanial effect.
  • Potent and orally active xanthine oxidase inhibitor
  • Can be used in the research of hyperuricemia and gout
  • Decreases the expression of HIF-1α and HIF-2α protein
  • Reduces angiogenesis traits of HUVEC cells
  • Exhibits anti-depressant activity in mouse models
  • Induces anti-nociception activity in mouse models
  • Shows anti-leishmanial effect
Catalog No. 50-004-05514