Learn More
Medchemexpress LLC Allopurinol | 315-30-0 | 599413 | 100.0% | 25 G
SDP
Catalog No. 5000406146
Encompass_Preferred
Allopurinol is a potent and orally active xanthine oxidase inhibitor. It is utilized in the research of hyperuricemia and gout, and also demonstrates anti-depressant, anti-nociception, and anti-leishmanial activities by decreasing the expression of HIF-1α and HIF-2α protein.
  • Potent and orally active xanthine oxidase inhibitor
  • Used in hyperuricemia and gout research
  • Decreases expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant activity
  • Exhibits anti-nociception activity
  • Exhibits anti-leishmanial activity
Catalog No. 50-004-06146 Supplier Medchemexpress LLC Supplier No. 25GHYB0219
Allopurinol is a potent and orally active xanthine oxidase inhibitor. It is utilized in the research of hyperuricemia and gout, and also demonstrates anti-depressant, anti-nociception, and anti-leishmanial activities by decreasing the expression of HIF-1α and HIF-2α protein.
  • Potent and orally active xanthine oxidase inhibitor
  • Used in hyperuricemia and gout research
  • Decreases expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant activity
  • Exhibits anti-nociception activity
  • Exhibits anti-leishmanial activity