Learn More
Medchemexpress LLC Allopurinol | 315-30-0 | 99.9% | 100 G
SDP
Catalog No. 5000407231
Encompass_Preferred
Allopurinol is a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. It is primarily used in the research of hyperuricemia and gout. Additionally, Allopurinol has been observed to decrease the expression of HIF-1α and HIF-2α protein, and exhibits anti-depressant, anti-nociception, and anti-leishmanial effects.
  • Potent and orally active xanthine oxidase inhibitor
  • Used in hyperuricemia and gout research
  • Decreases HIF-1α and HIF-2α protein expression
  • Exhibits anti-depressant activity
  • Shows anti-nociception activity
  • Demonstrates anti-leishmanial effects
Catalog No. 50-004-07231 Supplier Medchemexpress LLC Supplier No. HYB0219100G
Allopurinol is a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. It is primarily used in the research of hyperuricemia and gout. Additionally, Allopurinol has been observed to decrease the expression of HIF-1α and HIF-2α protein, and exhibits anti-depressant, anti-nociception, and anti-leishmanial effects.
  • Potent and orally active xanthine oxidase inhibitor
  • Used in hyperuricemia and gout research
  • Decreases HIF-1α and HIF-2α protein expression
  • Exhibits anti-depressant activity
  • Shows anti-nociception activity
  • Demonstrates anti-leishmanial effects