Learn More
Medchemexpress LLC Allopurinol | 315-30-0 | 99.9% | 5 G
SDP
Catalog No. 5000405521
Encompass_Preferred
Allopurinol is a potent and orally active xanthine oxidase inhibitor used in the research of hyperuricemia and gout. It decreases the expression of HIF-1α and HIF-2α protein. Allopurinol also exhibits anti-depressant, anti-nociception, and anti-leishmanial effects.
  • Potent and orally active xanthine oxidase inhibitor
  • Used in research of hyperuricemia and gout
  • Decreases expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant activity
  • Shows anti-nociception activity
  • Possesses anti-leishmanial effects
Catalog No. 50-004-05521 Supplier Medchemexpress LLC Supplier No. HYB02195G
Allopurinol is a potent and orally active xanthine oxidase inhibitor used in the research of hyperuricemia and gout. It decreases the expression of HIF-1α and HIF-2α protein. Allopurinol also exhibits anti-depressant, anti-nociception, and anti-leishmanial effects.
  • Potent and orally active xanthine oxidase inhibitor
  • Used in research of hyperuricemia and gout
  • Decreases expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant activity
  • Shows anti-nociception activity
  • Possesses anti-leishmanial effects