Learn More
Medchemexpress LLC Allopurinol | 315-30-0 | 99.96% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB021910G
Encompass_Preferred
Allopurinol is a potent, orally active xanthine oxidase inhibitor used in research for conditions like hyperuricemia and gout. It decreases the expression of HIF-1α and HIF-2α proteins and exhibits anti-depressant, anti-nociception, and anti-leishmanial activities. Its IC50 value ranges from 0.2-50 μM.
  • Potent xanthine oxidase inhibitor
  • Useful in hyperuricemia and gout research
  • Decreases HIF-1α and HIF-2α protein expression
  • Exhibits anti-depressant activity
  • Shows anti-nociception activity
  • Demonstrates anti-leishmanial effects
Catalog No. 50-004-05676