Learn More
Medchemexpress LLC Allopurinol | 315-30-0 | 99.96% | 50 G
SDP
Catalog No. 5000406138
Encompass_Preferred
Allopurinol is a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. It can be used in the research of hyperuricemia and gout. Allopurinol decreases the expression of HIF-1α and HIF-2α protein. It also shows anti-depressant and anti-nociception activity, and an anti-leishmanial effect.
  • Potent and orally active xanthine oxidase inhibitor
  • IC50 value of 0.2-50 μM
  • Used in research for hyperuricemia and gout
  • Decreases the expression of HIF-1α and HIF-2α protein
  • Shows anti-depressant activity
  • Shows anti-nociception activity
  • Exhibits anti-leishmanial effect
Catalog No. 50-004-06138 Supplier Medchemexpress LLC Supplier No. HYB021950G
Allopurinol is a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. It can be used in the research of hyperuricemia and gout. Allopurinol decreases the expression of HIF-1α and HIF-2α protein. It also shows anti-depressant and anti-nociception activity, and an anti-leishmanial effect.
  • Potent and orally active xanthine oxidase inhibitor
  • IC50 value of 0.2-50 μM
  • Used in research for hyperuricemia and gout
  • Decreases the expression of HIF-1α and HIF-2α protein
  • Shows anti-depressant activity
  • Shows anti-nociception activity
  • Exhibits anti-leishmanial effect