Learn More
Alkali Scientific Allopurinol [4 Hydroxypyrazolo [3,4 d]pyrimidine], 100 G
SDP
Catalog No. 5000660741
Encompass_Preferred
Amikacin Disulfate is an aminoglycoside antibiotic commonly used in laboratory research for its broad spectrum antibacterial activity against Gram negative and certain Gram positive organisms. It functions by binding to the bacterial 30S ribosomal subunit, disrupting protein synthesis and leading to cell death. Due to its stability against many bacterial resistance enzymes, it is frequently used in microbiological studies and culture systems where reliable antibacterial control is required. This compound is widely applied in cell culture contamination prevention, antimicrobial susceptibility testing, and bacterial inhibition studies. Supplied in a 25 gram quantity, it is suitable for routine laboratory use and bulk preparation of working solutions.
Catalog No. 50-006-60741 Supplier Alkali Scientific Supplier No. ALL002CP
Amikacin Disulfate is an aminoglycoside antibiotic commonly used in laboratory research for its broad spectrum antibacterial activity against Gram negative and certain Gram positive organisms. It functions by binding to the bacterial 30S ribosomal subunit, disrupting protein synthesis and leading to cell death. Due to its stability against many bacterial resistance enzymes, it is frequently used in microbiological studies and culture systems where reliable antibacterial control is required. This compound is widely applied in cell culture contamination prevention, antimicrobial susceptibility testing, and bacterial inhibition studies. Supplied in a 25 gram quantity, it is suitable for routine laboratory use and bulk preparation of working solutions.