Learn More
Alkali Scientific Allopurinol [4 Hydroxypyrazolo [3,4 d]pyrimidine], 25 G
SDP
Catalog No. 5000660434
Encompass_Preferred
Allopurinol is a xanthine oxidase inhibitor used in biochemical research to investigate purine metabolism and enzyme inhibition pathways. It prevents the formation of uric acid by blocking the activity of xanthine oxidase, allowing researchers to study metabolic regulation and oxidative processes in biological systems. The compound is commonly applied in pharmacological studies, enzymatic assays, and cellular research focused on metabolic disorders and oxidative stress. Its predictable inhibitory mechanism makes it useful for controlled experimental conditions. Supplied in a 25 gram quantity, it is suitable for routine laboratory use and preparation of research solutions.
Catalog No. 50-006-60434 Supplier Alkali Scientific Supplier No. ALL004CP
Allopurinol is a xanthine oxidase inhibitor used in biochemical research to investigate purine metabolism and enzyme inhibition pathways. It prevents the formation of uric acid by blocking the activity of xanthine oxidase, allowing researchers to study metabolic regulation and oxidative processes in biological systems. The compound is commonly applied in pharmacological studies, enzymatic assays, and cellular research focused on metabolic disorders and oxidative stress. Its predictable inhibitory mechanism makes it useful for controlled experimental conditions. Supplied in a 25 gram quantity, it is suitable for routine laboratory use and preparation of research solutions.