Learn More
Alkali Scientific Allopurinol [4 Hydroxypyrazolo [3,4 d]pyrimidine], 5 G
SDP
Catalog No. 5000660854
Encompass_Preferred
Allopurinol is a xanthine oxidase inhibitor used in laboratory research to study purine metabolism and enzyme activity. It blocks the conversion of xanthine into uric acid, making it useful for investigating metabolic pathways and oxidative stress responses. Researchers commonly use this compound in biochemical assays and pharmacological studies focused on enzyme inhibition and cellular metabolism. Its consistent activity allows for controlled experimental conditions in laboratory applications. Supplied in a 5 gram quantity, it is suitable for small scale research experiments and analytical procedures requiring precise measurements.
Catalog No. 50-006-60854 Supplier Alkali Scientific Supplier No. ALL005CP
Allopurinol is a xanthine oxidase inhibitor used in laboratory research to study purine metabolism and enzyme activity. It blocks the conversion of xanthine into uric acid, making it useful for investigating metabolic pathways and oxidative stress responses. Researchers commonly use this compound in biochemical assays and pharmacological studies focused on enzyme inhibition and cellular metabolism. Its consistent activity allows for controlled experimental conditions in laboratory applications. Supplied in a 5 gram quantity, it is suitable for small scale research experiments and analytical procedures requiring precise measurements.