Learn More
Selleck Chemical LLC Allopurinol S1630-100mg
SDP
Catalog No. 503110684
Encompass_Preferred
Allopurinol is a purine analog inhibitor of the enzyme xanthine oxidase used to treat gout or kidney stones and to decrease levels of uric acid
Catalog No. 50-311-0684 Supplier Selleck Chemical LLC Supplier No. S1630100MG
Allopurinol is a purine analog inhibitor of the enzyme xanthine oxidase used to treat gout or kidney stones and to decrease levels of uric acid