Learn More
Medchemexpress LLC Allopurinol-d2 | 916979-34-5 | 99.4% | 1 MG
SDP
Catalog No. 5000405639
Encompass_Preferred
Allopurinol-d2 is a deuterium-labeled version of Allopurinol, which is a potent xanthine oxidase inhibitor (IC50 values of 0.2 to 50 μM). It is utilized in research for hyperuricemia, gout, and exhibits antileishmanial effects. Deuterium substitution has been explored for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Potent xanthine oxidase inhibitor
  • Used for research of hyperuricemia and gout
  • Exhibits antileishmanial effect
  • Can be used as a tracer for quantitation
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05639 Supplier Medchemexpress LLC Supplier No. HYB0219S1MG
Allopurinol-d2 is a deuterium-labeled version of Allopurinol, which is a potent xanthine oxidase inhibitor (IC50 values of 0.2 to 50 μM). It is utilized in research for hyperuricemia, gout, and exhibits antileishmanial effects. Deuterium substitution has been explored for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Potent xanthine oxidase inhibitor
  • Used for research of hyperuricemia and gout
  • Exhibits antileishmanial effect
  • Can be used as a tracer for quantitation
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS