Learn More
Cayman Chemical Allopurinol Inhibitors
SDP
Catalog No. 502888896
Encompass_Preferred
An isomer of hypoxanthine that inhibits xanthine oxidoreductase (IC50 values between 0.2 and 50 μM); lowers serum and urinary uric acid levels in vivo and is used to treat gout and hyperuricemia
Catalog No. 50-288-8896 Supplier Cayman Chemical Supplier No. 1001259725 G
An isomer of hypoxanthine that inhibits xanthine oxidoreductase (IC50 values between 0.2 and 50 μM); lowers serum and urinary uric acid levels in vivo and is used to treat gout and hyperuricemia