Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allopurinol Related Compound E Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013890 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3890 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3890 Supplier MilliporeSigma Supelco Supplier No. PHR200950MG
Add to Cart
Add to Cart

Specifications

CAS 31055-19-3
Linear Formula C7H9N3O3
Synonym Ethyl 3-(formylamino)-1 H-pyrazole-4-carboxylate; Ethyl 5-(formylamino)-1 H-pyrazole-4-carboxylate
Type Certified Reference Material
Quantity 50 mg
Formula Weight 183.16
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.