Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Allopurinol riboside | 16220-07-8 | 99.1% | 268.23 g/mol | C10H12N4O5 | 10 MG
SDP

Catalog No. 5000212467
Encompass_Preferred

Allopurinol riboside is a drug metabolite of allopurinol reported to have activity against parasites. It is provided for research use as both solid material and concentrated DMSO solutions, with documented solubility, formulation, and storage guidance to support in vitro and in vivo studies.

  • Drug metabolite with reported antiparasitic activity.
  • Molecular formula C10H12N4O5; molecular weight 268.23 g/mol.
  • High purity (≈99%).
  • Available as solid (1 mg, 5 mg, 10 mg) and as 10 mM solution in DMSO.
  • Solubility: DMSO 100 mg/mL; stock preparation guidance for 1-10 mM.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Formulation guidance provided for achieving ≥2.5 mg/mL in several in vivo vehicles.

Catalog No. 50-002-12467 Supplier Medchemexpress LLC Supplier No. HY10139710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allopurinol riboside is a drug metabolite of allopurinol reported to have activity against parasites. It is provided for research use as both solid material and concentrated DMSO solutions, with documented solubility, formulation, and storage guidance to support in vitro and in vivo studies.

  • Drug metabolite with reported antiparasitic activity.
  • Molecular formula C10H12N4O5; molecular weight 268.23 g/mol.
  • High purity (≈99%).
  • Available as solid (1 mg, 5 mg, 10 mg) and as 10 mM solution in DMSO.
  • Solubility: DMSO 100 mg/mL; stock preparation guidance for 1-10 mM.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Formulation guidance provided for achieving ≥2.5 mg/mL in several in vivo vehicles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.