Learn More
Selleck Chemical LLC Allopurinol Sodium S1631-1g
SDP
Catalog No. 503108685
Encompass_Preferred
Allopurinol Sodium is a xanthine oxidase inhibitor with an IC50 of 7 82 0 12 M
Catalog No. 50-310-8685 Supplier Selleck Chemical LLC Supplier No. S16311G
Allopurinol Sodium is a xanthine oxidase inhibitor with an IC50 of 7 82 0 12 M