Learn More
Selleck Chemical LLC Allopurinol Sodium S1631-25mg
SDP
Catalog No. 503110126
Encompass_Preferred
Allopurinol Sodium is a xanthine oxidase inhibitor with an IC50 of 7 82 0 12 M
Catalog No. 50-311-0126 Supplier Selleck Chemical LLC Supplier No. S163125MG
Allopurinol Sodium is a xanthine oxidase inhibitor with an IC50 of 7 82 0 12 M