Learn More
Medchemexpress LLC Allopurinol (sodium) | 17795-21-0 | 99.9% | 25 MG
SDP
Catalog No. 5000451023
Encompass_Preferred
Allopurinol sodium is a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. It is primarily used in the research of hyperuricemia and gout. This compound decreases the expression of HIF-1α and HIF-2α protein, and demonstrates anti-depressant, anti-nociception, and anti-leishmanial effects.
  • Potent and orally active xanthine oxidase inhibitor
  • Decreases the expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant activity
  • Demonstrates anti-nociception activity
  • Shows anti-leishmanial effects
Catalog No. 50-004-51023 Supplier Medchemexpress LLC Supplier No. 25MGHYB0219A
Allopurinol sodium is a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. It is primarily used in the research of hyperuricemia and gout. This compound decreases the expression of HIF-1α and HIF-2α protein, and demonstrates anti-depressant, anti-nociception, and anti-leishmanial effects.
  • Potent and orally active xanthine oxidase inhibitor
  • Decreases the expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant activity
  • Demonstrates anti-nociception activity
  • Shows anti-leishmanial effects