Learn More
Medchemexpress LLC Allopurinol sodium | 17795-21-0 | 99.90% | 100 MG
SDP
Catalog No. 5000405788
Encompass_Preferred
Allopurinol sodium is a potent and orally active xanthine oxidase inhibitor (IC₅₀ value of 0.2-50 μM) used in research for hyperuricemia and gout. It decreases the expression of HIF-1α and HIF-2α protein, and also demonstrates anti-depressant, anti-nociception, and anti-leishmanial effects.
  • Potent and orally active xanthine oxidase inhibitor
  • Research applications include hyperuricemia and gout
  • Decreases expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant and anti-nociception activity
  • Shows anti-leishmanial effect
  • Reduces angiogenesis traits in HUVEC cells
  • Reduces elevated IL-6 levels and phosphorylation of JAK2 and STAT3
  • Normalizes mitochondrial damage in cardiomyocytes
  • Upregulates HPRT, XOR, PNP, Nrf2, SOD-1, and CAT genes
Catalog No. 50-004-05788 Supplier Medchemexpress LLC Supplier No. HYB0219A100MG
Allopurinol sodium is a potent and orally active xanthine oxidase inhibitor (IC₅₀ value of 0.2-50 μM) used in research for hyperuricemia and gout. It decreases the expression of HIF-1α and HIF-2α protein, and also demonstrates anti-depressant, anti-nociception, and anti-leishmanial effects.
  • Potent and orally active xanthine oxidase inhibitor
  • Research applications include hyperuricemia and gout
  • Decreases expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant and anti-nociception activity
  • Shows anti-leishmanial effect
  • Reduces angiogenesis traits in HUVEC cells
  • Reduces elevated IL-6 levels and phosphorylation of JAK2 and STAT3
  • Normalizes mitochondrial damage in cardiomyocytes
  • Upregulates HPRT, XOR, PNP, Nrf2, SOD-1, and CAT genes