Learn More
TARGETMOL CHEMICALS INC Allopurinol Sodium 50MG
SDP
Catalog No. 502880044
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.Allopurinol Sodium is a xanthine oxidase inhibitor with an IC50 of 7.82±0.12 uM. Purity 99.92%
Catalog No. 50-288-0044 Supplier TARGETMOL CHEMICALS INC Supplier No. T637650MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.Allopurinol Sodium is a xanthine oxidase inhibitor with an IC50 of 7.82±0.12 uM. Purity 99.92%