Learn More
Medchemexpress LLC Allopurinol (standard) | 315-30-0 | 100.0% | 100 MG
SDP
Catalog No. 5000405692
Encompass_Preferred
Allopurinol (Standard) is the analytical standard of Allopurinol, intended for research and analytical applications. It is a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. This compound is used in the research of hyperuricemia and gout. It decreases the expression of HIF-1α and HIF-2α protein, and exhibits anti-depressant, anti-nociception, and anti-leishmanial activities. As an analytical standard, it serves as a reference for qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Potent and orally active xanthine oxidase inhibitor
  • Supports research on hyperuricemia and gout
  • Decreases expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant, anti-nociception, and anti-leishmanial activities
  • Reference standard for assay in HPLC, GC, and MS experiments
Catalog No. 50-004-05692 Supplier Medchemexpress LLC Supplier No. HYB0219R100MG
Allopurinol (Standard) is the analytical standard of Allopurinol, intended for research and analytical applications. It is a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. This compound is used in the research of hyperuricemia and gout. It decreases the expression of HIF-1α and HIF-2α protein, and exhibits anti-depressant, anti-nociception, and anti-leishmanial activities. As an analytical standard, it serves as a reference for qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Potent and orally active xanthine oxidase inhibitor
  • Supports research on hyperuricemia and gout
  • Decreases expression of HIF-1α and HIF-2α protein
  • Exhibits anti-depressant, anti-nociception, and anti-leishmanial activities
  • Reference standard for assay in HPLC, GC, and MS experiments