Learn More
Medchemexpress LLC Allopurinol (Standard) | 315-30-0 | 100.0% | 250 MG
SDP
Catalog No. 5000406821
Encompass_Preferred
Allopurinol (Standard) is an analytical standard intended for research and analytical applications. It is a potent and orally active xanthine oxidase inhibitor, with an IC50 value of 0.2-50 μM, making it suitable for research into hyperuricemia and gout. The compound can be used as a reference standard in techniques such as HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Functions as a potent and orally active xanthine oxidase inhibitor
  • Used in the research of hyperuricemia and gout
  • Decreases the expression of HIF-1α and HIF-2α protein
  • Shows anti-depressant and anti-nociception activity
  • Exhibits anti-leishmanial effect
Catalog No. 50-004-06821 Supplier Medchemexpress LLC Supplier No. 250MGHYB0219R
Allopurinol (Standard) is an analytical standard intended for research and analytical applications. It is a potent and orally active xanthine oxidase inhibitor, with an IC50 value of 0.2-50 μM, making it suitable for research into hyperuricemia and gout. The compound can be used as a reference standard in techniques such as HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Functions as a potent and orally active xanthine oxidase inhibitor
  • Used in the research of hyperuricemia and gout
  • Decreases the expression of HIF-1α and HIF-2α protein
  • Shows anti-depressant and anti-nociception activity
  • Exhibits anti-leishmanial effect