Learn More
Medchemexpress LLC Allopurinol (Standard) | 315-30-0 | 599413 | 99.97% | 25 MG
SDP
Catalog No. 5000450982
Encompass_Preferred
Allopurinol (Standard) is the analytical standard of Allopurinol, intended for research and analytical applications. It is a potent and orally active xanthine oxidase inhibitor, utilized in the research of hyperuricemia and gout. It also decreases the expression of HIF-1α and HIF-2α proteins, and exhibits anti-depressant, anti-nociception, and anti-leishmanial activities.
  • Potent and orally active xanthine oxidase inhibitor
  • Used in research of hyperuricemia and gout
  • Decreases expression of HIF-1α and HIF-2α protein
  • Shows anti-depressant and anti-nociception activity
  • Exhibits anti-leishmanial effect
Catalog No. 50-004-50982 Supplier Medchemexpress LLC Supplier No. 25MGHYB0219R
Add to Cart
Add to Cart
Allopurinol (Standard) is the analytical standard of Allopurinol, intended for research and analytical applications. It is a potent and orally active xanthine oxidase inhibitor, utilized in the research of hyperuricemia and gout. It also decreases the expression of HIF-1α and HIF-2α proteins, and exhibits anti-depressant, anti-nociception, and anti-leishmanial activities.
  • Potent and orally active xanthine oxidase inhibitor
  • Used in research of hyperuricemia and gout
  • Decreases expression of HIF-1α and HIF-2α protein
  • Shows anti-depressant and anti-nociception activity
  • Exhibits anti-leishmanial effect