Learn More
Medchemexpress LLC Allopurinol (Standard) | 315-30-0 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0219R500MG
Encompass_Preferred
Allopurinol (Standard) is the analytical standard of Allopurinol, a potent and orally active xanthine oxidase inhibitor with an IC50 value of 0.2-50 μM. It is intended for research and analytical applications, particularly in studies of hyperuricemia and gout, and is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. Allopurinol also demonstrates anti-depressant, anti-nociception, and anti-leishmanial activity by decreasing the expression of HIF-1α and HIF-2α protein.
  • Analytical standard for research and analytical applications
  • Potent and orally active xanthine oxidase inhibitor
  • Used in research of hyperuricemia and gout
  • Decreases expression of HIF-1α and HIF-2α protein
  • Shows anti-depressant and anti-nociception activity
  • Exhibits anti-leishmanial effect
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-05776