Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Alloy 8 Metric Precision Weight Sets, Class 3

Catalog No. 0221520B Shop All Troemner Products
Change view
Click to view available options
Material:
Stainless Steel
Quantity:
17/Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Quantity
02-215-20B Stainless Steel 17/Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-215-20B Supplier Troemner™ Supplier No. 80859289
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Forceps

Excellent for high-precision calibrations and reference standards

Manufactured from a specially developed austenitic stainless-steel—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless-steel.

  • Each fractional weight is embossed with its value
  • Weights from 1 to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
  • Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
  • Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
  • Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with 2 (2mg, 20mg, etc.) and one weight for all other inclusive values
TRUSTED_SUSTAINABILITY

Specifications

Product Type Medium Metric Analytical Weight Set
Classification ASTM Class 3
Material Stainless Steel
Weight (Metric) 10 mg to 100 g
Mass 10 mg to 100 g
Includes Cylindrical weight: 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, Polycarbonate case
Quantity 17/Set
Packaging Quantity Multiple
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.