Learn More
MSC Columbus McKinnon Alloy Anchor Shackle, 5/8 in, Screw Pin, 5 ton
Catalog No. 5000514080
Encompass
The Columbus McKinnon Alloy Anchor Shackle is a robust and reliable lifting component designed for demanding applications. Manufactured from domestically produced alloy steel, it features a screw pin for secure attachment and a 5 ton working load limit, making it suitable for various industrial and marine uses.
  • Alloy steel construction ensures durability and strength.
  • Screw pin design provides secure and easy attachment.
  • 5/8 inch size accommodates a range of rigging needs.
  • Working load limit of 5 tons (10,000 lbs) for heavy-duty lifting.
  • Self-colored finish offers a natural appearance.
  • Design factor of 5:1 provides enhanced safety margins.
  • Deforms significantly before failure, offering a visual warning.
  • Permanently marked with working load limit and traceability codes.
  • Meets ASME B30.26 and RR-C-271 standards for quality and performance.
Catalog No. 50-005-14080 Supplier MSC Supplier No. 67788703
The Columbus McKinnon Alloy Anchor Shackle is a robust and reliable lifting component designed for demanding applications. Manufactured from domestically produced alloy steel, it features a screw pin for secure attachment and a 5 ton working load limit, making it suitable for various industrial and marine uses.
  • Alloy steel construction ensures durability and strength.
  • Screw pin design provides secure and easy attachment.
  • 5/8 inch size accommodates a range of rigging needs.
  • Working load limit of 5 tons (10,000 lbs) for heavy-duty lifting.
  • Self-colored finish offers a natural appearance.
  • Design factor of 5:1 provides enhanced safety margins.
  • Deforms significantly before failure, offering a visual warning.
  • Permanently marked with working load limit and traceability codes.
  • Meets ASME B30.26 and RR-C-271 standards for quality and performance.