Learn More
MSC Campbell Swivel Hoist Hook, Alloy Steel, 3 Ton Capacity, 1 ea
Catalog No. 5000512577
Encompass
This swivel hoist hook is drop-forged from durable alloy steel, offering a high-strength solution for rigging and lifting applications. Designed for secure attachment, the hook features an integrated latch to help retain slings and rigging components. It is painted orange and manufactured in the USA, meeting the requirements of ASME B30.10.
  • Capacity of 3 tons (6,000 lb)
  • Design factor of 5 to 1
  • Overall length of 8-1/4 in. and width of 4-1/8 in.
  • Hook opening of 1-5/32 in.
  • Includes a heavy-gauge high strength steel latch with yellow chromate finish
  • Integrated forged bosses for added security
  • Latch marking forged directly into the hook for identification
Catalog No. 50-005-12577 Supplier MSC Supplier No. 50538701
This swivel hoist hook is drop-forged from durable alloy steel, offering a high-strength solution for rigging and lifting applications. Designed for secure attachment, the hook features an integrated latch to help retain slings and rigging components. It is painted orange and manufactured in the USA, meeting the requirements of ASME B30.10.
  • Capacity of 3 tons (6,000 lb)
  • Design factor of 5 to 1
  • Overall length of 8-1/4 in. and width of 4-1/8 in.
  • Hook opening of 1-5/32 in.
  • Includes a heavy-gauge high strength steel latch with yellow chromate finish
  • Integrated forged bosses for added security
  • Latch marking forged directly into the hook for identification