Learn More
VWR INTERNATIONAL INC ALLUM. WEIGH DISH 200ML / 50PK
Catalog No. NC9047004
Encompass_Preferred
Disposable Aluminum Weighing Dishes, Capacity 200 mL (6.8 oz.), I.D x D 11.0 x 2.9 cm (45/16 x 11/8 in.)
Catalog No. NC9047004 Supplier VWR INTERNATIONAL INC Supplier No. 25433022
Disposable Aluminum Weighing Dishes, Capacity 200 mL (6.8 oz.), I.D x D 11.0 x 2.9 cm (45/16 x 11/8 in.)