Learn More
Medchemexpress LLC Allura red AC | 25956-17-6 | MFCD00059526 | 99.0% | 496.42 g/mol | C18H14N2Na2O8S2 | 100MG
SDP
Supplier:  Medchemexpress LLC HY123630100MG
Encompass_Preferred
Allura Red AC is a water-soluble red food colorant supplied as a deep red powder for research and analytical applications. It is used in studies of food additive safety, intestinal barrier function, and inflammatory pathways. The material dissolves readily in water and DMSO and is provided in small, high-purity quantities suitable for laboratory use.
  • High purity of 99.0%.
  • Provided as a deep red, water-soluble powder.
  • Good solubility: H2O 100 mg/mL, DMSO 62.5 mg/mL (may require ultrasonic).
  • Compact 100 mg pack for small-scale experiments.
  • Stable when stored sealed away from moisture and light; solvent storage guidelines available.
Catalog No. 50-002-36706