Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl 2,2,2-Trichloroacetimidate 98.0+%, TCI America™
SDP

Catalog No. A21865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21865G Supplier TCI America Supplier No. A21865G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51479-73-3
Molecular Formula C5H6Cl3NO
Molecular Weight (g/mol) 202.459
MDL Number MFCD00190710
InChI Key UJYZRNWTLPBNOR-UHFFFAOYSA-N
Synonym 2,2,2-Trichloroacetimidic Acid Allyl Ester
PubChem CID 316905
IUPAC Name prop-2-enyl 2,2,2-trichloroethanimidate
SMILES C=CCOC(=N)C(Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 74°C
Quantity 5 g
Formula Weight 202.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl 2,2,2-Trichloroacetimidate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.