Learn More
Medchemexpress LLC Allyl 2-aminoethyl 10g
SDP
Catalog No. 5000205168
Encompass_Preferred
Allyl (2-aminoethyl)carbamate hydrochloride is a cleavable linker
Catalog No. 50-002-05168 Supplier Medchemexpress LLC Supplier No. HYW09876410G
Allyl (2-aminoethyl)carbamate hydrochloride is a cleavable linker