Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Allyl (2-aminoethyl) carbamate | 223741-66-0 | 98.0% | C6H12N2O2 | 25 MG
SDP

Catalog No. 5000356568
Encompass_Preferred

Allyl (2-aminoethyl)carbamate is a cleavable linker, primarily used as a laboratory chemical for the manufacture of substances. It appears as a colorless to light yellow liquid.

  • Molecular weight: 144.17
  • Boiling point: 248.6±33.0 °C at 760 mmHg
  • Flash point: 104.1°C
  • Relative density: 1.039 g/cm³
  • Recommended storage: 4°C for sealed storage, away from moisture and light
  • For storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture and light)
  • Shipping conditions: Room temperature if less than 2 weeks
  • For research use only

Catalog No. 50-003-56568 Supplier Medchemexpress LLC Supplier No. 25MGHY145735
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allyl (2-aminoethyl)carbamate is a cleavable linker, primarily used as a laboratory chemical for the manufacture of substances. It appears as a colorless to light yellow liquid.

  • Molecular weight: 144.17
  • Boiling point: 248.6±33.0 °C at 760 mmHg
  • Flash point: 104.1°C
  • Relative density: 1.039 g/cm³
  • Recommended storage: 4°C for sealed storage, away from moisture and light
  • For storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture and light)
  • Shipping conditions: Room temperature if less than 2 weeks
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.