Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl [3-(2-Nitrobenzenesulfonamido)propyl]carbamate 98.0+%, TCI America™
SDP

Catalog No. N07171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N07171G Supplier TCI America Supplier No. N07171G

Chemical Identifiers

CAS 312283-45-7
Molecular Formula C13H17N3O6S
Molecular Weight (g/mol) 343.354
InChI Key INKIDQWOEVNHHA-UHFFFAOYSA-N
Synonym [3-(2-Nitrobenzenesulfonamido)propyl]carbamic Acid Allyl Ester
PubChem CID 10497545
IUPAC Name prop-2-enyl N-[3-[(2-nitrophenyl)sulfonylamino]propyl]carbamate
SMILES C=CCOC(=O)NCCCNS(=O)(=O)C1=CC=CC=C1[N+](=O)[O-]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 343.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Allyl [3-(2-Nitrobenzenesulfonamido)propyl]carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.