Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allyl Bromide

Catalog No. A29585500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
3 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29585500G 500 g
A29585100G 100 g
A295851KG 1 kg
A295853KG 3 kg
A295855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A29585500G Supplier Sigma Aldrich Supplier No. A29585500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Allyl Bromide

Specifications

CAS 106-95-6
Melting Point -119°C
Density 1.398 g/mL at 25°C
Boiling Point 70°C to 71°C
Molecular Formula C3H5Br
Refractive Index n20/D 1.469
Linear Formula CH2=CHCH2Br
MDL Number MFCD00000244
Quantity 500 g
Synonym 3-Bromo-1-propene
Molecular Weight (g/mol) 120.98 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.