Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl(chloromethyl)dimethylsilane, 97+%, Thermo Scientific™

Catalog No. AA4195314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4195314 25 g
AA4195303 1 g
AA4195306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4195314 Supplier Thermo Scientific Chemicals Supplier No. 4195314

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75422-66-1
Molecular Formula C6H13ClSi
Molecular Weight (g/mol) 148.71
MDL Number MFCD00012045
InChI Key UHHLOVCFFWGSMO-UHFFFAOYSA-N
Synonym allyl chloromethyl dimethylsilane, silane, chloromethyl dimethyl-2-propenyl, chloromethyl dimethylprop-2-en-1-ylsilane, acmc-1bfca, dimethylallylchloromethylsilane, 3-chloromethyldimethylsilyl-1-propene, allyl chloromethyl dimethyl dimethylsilane, silane,allyl, chloromethyl ,dimethyl, chloromethyl dimethyl prop-2-en-1-yl silane
PubChem CID 556526
IUPAC Name chloromethyl-dimethyl-prop-2-enylsilane
SMILES C[Si](C)(CCl)CC=C

Specifications

Boiling Point 165°C (50mmHg)
Physical Form Liquid
Quantity 25 g
UN Number UN1993
Flash Point 27°C (81°F)
Formula Weight 148.71
Percent Purity ≥97%
Refractive Index 1.449
Sensitivity Moisture sensitive
Density 0.907 g/mL
Chemical Name or Material Allyl(chloromethyl)dimethylsilane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.