Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Cyanide 98%, Thermo Scientific™

Catalog No. AC153705000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153705000 500 mL
AC153701000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153705000 Supplier Thermo Scientific Chemicals Supplier No. 153705000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-75-1
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.091
InChI Key SJNALLRHIVGIBI-UHFFFAOYSA-N
Synonym 3-butenenitrile, allyl cyanide, 3-butenylnitrile, 3-butenonitrile, allylnitrile, vinylacetonitrile, 3-cyanopropene, 3-cyano-1-propene, 1-butene-4-nitrile, propene-3-nitrile
PubChem CID 8009
IUPAC Name but-3-enenitrile
SMILES C=CCC#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Linear Formula H2C=CHCH2CN
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
Quantity 500 mL
Beilstein 02, 408
Merck Index 15, 281
Formula Weight 67.09
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Allyl cyanide, 98%
Show More Show Less
EINECSNumber 203-701-1
RTECSNumber EM8050000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.