Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Cyanide, MP Biomedicals

Catalog No. ICN203455 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203455 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203455 Supplier MP Biomedicals, Inc Supplier No. 0520345580

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109-75-1
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.091
InChI Key SJNALLRHIVGIBI-UHFFFAOYSA-N
Synonym 3-butenenitrile, allyl cyanide, 3-butenylnitrile, 3-butenonitrile, allylnitrile, vinylacetonitrile, 3-cyanopropene, 3-cyano-1-propene, 1-butene-4-nitrile, propene-3-nitrile
PubChem CID 8009
IUPAC Name but-3-enenitrile
SMILES C=CCC#N

Specifications

Quantity 100 g
Formula Weight 67.09
Chemical Name or Material Allyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.