Learn More
Strem, An Ascensus Company CAS# 1271-03-0. 500mg. Allyl(cyclopentadienyl)palladium(II), 98%.
SDP
Supplier:  Strem, An Ascensus Company 460065500MG
Encompass
CAS# 1271-03-0. 500mg. Allyl(cyclopentadienyl)palladium(II), 98%. . Molecular Weight: 212.58. Molecular Formula: C8H10Pd. Color/form: red solid Strem# 46-0065. http://www.strem.com/catalog/v/46-0065/
Catalog No. NC1094758