Learn More
Medchemexpress LLC Allyl α-D-galactopyranoside | 48149-72-0 | MFCD01320363 | 98.0% | 220.22 g·mol⁻¹ | C9H16O6 | 1 G
SDP
Catalog No. 5000300228
Encompass_Preferred
Allyl α-D-galactopyranoside is an allyl glycoside of D-galactopyranose used as a biochemical reagent and synthetic intermediate in glycobiology and carbohydrate chemistry. Supplied as a solid with high purity, it is commonly used in glycosylation studies, oligosaccharide assembly, and analytical research where a well-characterized carbohydrate building block is required.
  • High purity suitable for synthetic and analytical applications.
  • Solid powder form for easy handling and storage.
  • Stable under recommended storage conditions.
  • Useful building block for oligosaccharide synthesis.
  • Molecular formula C9H16O6 and molecular weight 220.22 g·mol⁻¹.
Catalog No. 50-003-00228 Supplier Medchemexpress LLC Supplier No. HYW0399151G
Allyl α-D-galactopyranoside is an allyl glycoside of D-galactopyranose used as a biochemical reagent and synthetic intermediate in glycobiology and carbohydrate chemistry. Supplied as a solid with high purity, it is commonly used in glycosylation studies, oligosaccharide assembly, and analytical research where a well-characterized carbohydrate building block is required.
  • High purity suitable for synthetic and analytical applications.
  • Solid powder form for easy handling and storage.
  • Stable under recommended storage conditions.
  • Useful building block for oligosaccharide synthesis.
  • Molecular formula C9H16O6 and molecular weight 220.22 g·mol⁻¹.