Learn More
Medchemexpress LLC Allyl α-D-galactopyranoside | 48149-72-0 | MFCD01320363 | 98.0% | 220.22 g/mol | C9H16O6 | 500 MG
SDP
Catalog No. 5000299497
Encompass_Preferred
Allyl α-D-galactopyranoside is an allyl glycoside used as a biochemical reagent and building block in glycobiology and carbohydrate chemistry. It is supplied as a white to off-white solid with high purity, suitable for synthetic and analytical applications.
  • High purity (98.0%) for reliable experimental results.
  • Solid form, white to off-white, easy to handle and store.
  • Molecular weight 220.22 g/mol and formula C9H16O6 for accurate stoichiometry.
  • Available in multiple package sizes, including 500 mg, for flexible use.
  • Suitable as a building block in oligosaccharide synthesis and glycobiology assays.
Catalog No. 50-002-99497 Supplier Medchemexpress LLC Supplier No. HYW039915500MG
Allyl α-D-galactopyranoside is an allyl glycoside used as a biochemical reagent and building block in glycobiology and carbohydrate chemistry. It is supplied as a white to off-white solid with high purity, suitable for synthetic and analytical applications.
  • High purity (98.0%) for reliable experimental results.
  • Solid form, white to off-white, easy to handle and store.
  • Molecular weight 220.22 g/mol and formula C9H16O6 for accurate stoichiometry.
  • Available in multiple package sizes, including 500 mg, for flexible use.
  • Suitable as a building block in oligosaccharide synthesis and glycobiology assays.