Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allyl glycidyl ether

Catalog No. A32608100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32608100ML 100 mL
A32608500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A32608100ML Supplier Sigma Aldrich Supplier No. A32608100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Allyl glycidyl ether (AGE) is a monomer that acts as a building block in the polymerization of poly(allyl glycidyl ether) by anionic ring opening mechanism. It is industrially prepared by the reacting allyl alcohol and epichlorohydrin.Useful in hydrosilylation, including the H2PtCl6-+xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.

Specifications

CAS 106-92-3
Density 0.962 g/mL (at 25°C)
Boiling Point 154°C
Molecular Formula C6H10O2
Refractive Index n20/D 1.433 (literature)
Linear Formula C6H10O2
Quantity 100 mL
MDL Number MFCD00005143
Synonym 1-Allyloxy-2,3-epoxypropane; Allyl 2,3-epoxypropyl ether
Molecular Weight (g/mol) 114.14
Percent Purity ≥99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.