Learn More
Sigma Aldrich Fine Chemicals Biosciences Allyl hexanoate 98 fCC fG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W2032034KGK
Encompass
Allyl hexanoate is a volatile flavoring compound with a pineapple flavor. It is naturally found in pineapple baked potatoes and mushrooms. Allyl hexanoate is used as a flavoring agent in chewing gum candies and baked goods.
Catalog No. 50-289-8347