Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allyl iodide

Catalog No. 23832525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-832-525G 25 g
23-832-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-832-525G Supplier Sigma Aldrich Supplier No. 23832525G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The interaction of allyl iodide with moo3, bi2o3 and molybdates of bi, co and mg has been studied in the temperature range of 270-480c.allyl iodide has been employed as:
  • Precursor for generation of allyl radicals to study their recombination in a single pulse shock tube with gas chromatographic measurements
  • Reagent used with allylindium sesquiiodide in the cis-double allylation of cyclopropenes

Specifications

CAS 556-56-9
Density 1.837 g/mL (at 25°C (literature))
Boiling Point 102°C to 103°C (lit.)
Molecular Formula C3H5I
Refractive Index n20/D 1.554 (literature)
Linear Formula CH2=CHCH2I
MDL Number MFCD00001094
Quantity 25 g
Synonym 3-Iodo-1-propene
Molecular Weight (g/mol) 167.98
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.