Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific Allyl isothiocyanate, 57-06-7, MFCD00004822, 25g

Catalog No. 5001413315
Encompass_Preferred

Molecular Formula C4H5NS, Purity 98%, Molecular Weight 99.16, Melting Point -80°, Boiling Point 148-154,

Catalog No. 50-014-13315 Supplier Matrix Scientific Supplier No. 05688125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C4H5NS, Purity 98%, Molecular Weight 99.16, Melting Point -80°, Boiling Point 148-154,

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.