Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Allyl methacrylate

Catalog No. 234931100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-493-1100ML 100 mL
23-493-1500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-493-1100ML Supplier Sigma Aldrich Supplier No. 234931100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Allyl methacrylate (AMA) is an acrylic monomer with two types of vinyl groups which include methacrylic and allylic based double bonds. It is mainly utilized as a cross-linking agent to increase the hardness of the base material. It also facilitates an improvement in the heat resistant property of the resin.

Specifications

CAS 96-05-9
Density 0.938 g/mL (at 25°C)
Boiling Point 59°C to 61°C (43 mmHg)
Molecular Formula C7H10O2
Linear Formula CH2=C(CH3)COOCH2CH=CH2
Refractive Index n20/D 1.436 (literature)
MDL Number MFCD00008592
Quantity 100 mL
Molecular Weight (g/mol) 126.15
Percent Purity 98%
Biological Activity Liver
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.