Learn More
Medchemexpress LLC HY-128447 100mg Medchemexpress, Allyl methyl sulfide CAS:10152-76-8 Purity:>98%
SDP
Catalog No. 501969749
Encompass_Preferred
Medchemexpress, HY-128447 100mg Allyl methyl sulfide CAS:10152-76-8 Allyl methyl sulfide is a bioactive organosulfur compound found in garlic. Allyl methyl sulfide exhibits antibacterial, antioxidant and anticancer properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9749 Supplier Medchemexpress LLC Supplier No. HY128447100MG
Medchemexpress, HY-128447 100mg Allyl methyl sulfide CAS:10152-76-8 Allyl methyl sulfide is a bioactive organosulfur compound found in garlic. Allyl methyl sulfide exhibits antibacterial, antioxidant and anticancer properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.